Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014842.2 | NP_001014842.1 | 3 | 1116 | ||
NM_001289006.1 | NP_001275935.1 | 3 | 910 | |||
NM_006405.6 | NP_006396.2 | 3 | 1116 | |||
GenBank mRNA | AK292110.1 | - | 3 | 805 | ||
AK304642.1 | - | 3 | 811 | |||
BC010856.2 | - | 2 | 725 | 3899246 | ||
BX161390.1 | - | 3 | 1036 | |||
BX161494.1 | - | 3 | 998 | |||
U94831.1 | - | 2 | 723 |