Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ugt1a6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145079.3 | NP_659545.2 | 297 | |||
GenBank mRNA | AK162514.1 | - | 293 | |||
AY227197.1 | - | 1 | 265 | |||
BC012716.1 | - | 291 | ||||
BC069940.1 | - | 287 | 5249697 | |||
D87867.1 | - | 1 | 169 | |||
U09930.1 | - | 1 | 179 | |||
U16818.1 | - | 297 |