Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SORBS1 SIRNA Assays ›
Gene Symbol:
SORBS1
Gene Name:
sorbin and SH3 domain containing 1
Gene Aliases:
CAP, FLAF2, R85FL, SH3D5, SH3P12, SORB1
Chromosome Location:
Chr.10: 95311773 - 95561420 on Build GRCh38
UniGene ID:
Hs.38621
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20769
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SORBS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034954.1
NP_001030126.1
16
1649
NM_001034955.1
NP_001030127.1
16
1715
NM_001034956.1
NP_001030128.1
14
1304
NM_001034957.1
NP_001030129.1
11
1019
NM_001290294.1
NP_001277223.1
16
1698
NM_001290295.1
NP_001277224.1
14
1233
NM_001290296.1
NP_001277225.1
14
1559
NM_001290297.1
NP_001277226.1
12
1046
NM_001290298.1
NP_001277227.1
14
1253
NM_006434.2
NP_006425.2
11
1019
NM_015385.3
NP_056200.1
15
1602
NM_024991.2
NP_079267.1
15
1329
XM_006717589.1
XP_006717652.1
16
1699
XM_006717593.1
XP_006717656.1
13
1184
XM_011539140.1
XP_011537442.1
18
2443
XM_011539150.1
XP_011537452.1
17
2368
XM_011539155.1
XP_011537457.1
15
2270
XM_011539167.1
XP_011537469.1
14
1553
XM_017015500.1
XP_016870989.1
17
2368
XM_017015501.1
XP_016870990.1
17
2368
XM_017015502.1
XP_016870991.1
18
2443
XM_017015503.1
XP_016870992.1
17
2368
XM_017015504.1
XP_016870993.1
16
1357
XM_017015505.1
XP_016870994.1
17
2368
XM_017015506.1
XP_016870995.1
15
1282
XM_017015507.1
XP_016870996.1
18
2443
XM_017015508.1
XP_016870997.1
16
1357
XM_017015509.1
XP_016870998.1
14
1211
XM_017015510.1
XP_016870999.1
17
2368
XM_017015511.1
XP_016871000.1
16
1357
XM_017015512.1
XP_016871001.1
13
1184
XM_017015513.1
XP_016871002.1
14
1259
XM_017015514.1
XP_016871003.1
14
1259
XM_017015515.1
XP_016871004.1
17
1726
XM_017015516.1
XP_016871005.1
16
1699
XM_017015517.1
XP_016871006.1
17
1726
XM_017015518.1
XP_016871007.1
16
1655
XM_017015519.1
XP_016871008.1
15
1628
XM_017015520.1
XP_016871009.1
16
1651
XM_017015521.1
XP_016871010.1
15
1580
XM_017015522.1
XP_016871011.1
15
1624
XM_017015523.1
XP_016871012.1
15
1580
XM_017015524.1
XP_016871013.1
14
1553
XM_017015525.1
XP_016871014.1
15
1286
XM_017015526.1
XP_016871015.1
16
1357
XM_017015527.1
XP_016871016.1
14
1211
XM_017015528.1
XP_016871017.1
15
1330
XM_017015529.1
XP_016871018.1
15
1282
XM_017015530.1
XP_016871019.1
16
1357
XM_017015531.1
XP_016871020.1
15
1286
XM_017015532.1
XP_016871021.1
14
1255
XM_017015533.1
XP_016871022.1
14
1259
XM_017015534.1
XP_016871023.1
16
1357
XM_017015535.1
XP_016871024.1
14
1211
XM_017015536.1
XP_016871025.1
15
1286
XM_017015537.1
XP_016871026.1
14
1255
XM_017015538.1
XP_016871027.1
15
1330
XM_017015539.1
XP_016871028.1
13
1184
XM_017015540.1
XP_016871029.1
14
1259
GenBank mRNA
AB037717.1
-
15
1261
AF136380.1
-
10
974
AF136381.1
-
13
1514
AF330623.1
-
11
1001
AJ489942.1
-
13
1208
AK294924.1
-
2
127
AK300407.1
-
14
1186
AK304130.1
-
2
190
AK304753.1
-
15
2245
AL117472.1
-
15
1560
AM260536.1
-
16
1656
BC042612.1
-
13
1626
BC152463.1
-
15
1261
8862599
BX640814.1
-
3
1264
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell-matrix adhesion
insulin receptor signaling pathway
positive regulation of signal transduction
glucose transport
cellular response to insulin stimulus
stress fiber assembly
positive regulation of glycogen biosynthetic process
positive regulation of glucose import
positive regulation of lipid biosynthetic process
focal adhesion assembly
positive regulation of establishment of protein localization to plasma membrane
actin binding
SH3/SH2 adaptor activity
insulin receptor binding
protein binding
cytoskeletal protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0