Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307939.1 | NP_001294868.1 | 2 | 428 | ||
NM_006465.3 | NP_006456.1 | 2 | 428 | |||
GenBank mRNA | AB208830.1 | - | 2 | 347 | ||
AF116846.1 | - | 2 | 360 | |||
AK298716.1 | - | 2 | 353 | |||
AK302034.1 | - | 2 | 428 | |||
BC041792.1 | - | 2 | 356 | 5297774 | ||
BC045545.1 | - | 2 | 402 | |||
BC060824.1 | - | 2 | 341 | |||
DB455943.1 | - | 2 | 418 | |||
DC396899.1 | - | 2 | 428 |