Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313972.1 | NP_001300901.1 | 2 | 339 | ||
NM_006472.5 | NP_006463.3 | 3 | 708 | |||
XM_017000085.1 | XP_016855574.1 | 3 | 657 | |||
GenBank mRNA | AK226090.1 | - | 3 | 620 | ||
AK301148.1 | - | 3 | 230 | |||
AK304670.1 | - | 2 | 339 | |||
AY594328.2 | - | 3 | 343 | |||
BC024305.1 | - | 3 | 237 | |||
BC093702.1 | - | 3 | 417 | 7939547 | ||
BC093704.1 | - | 3 | 417 | 7939549 | ||
S73591.1 | - | 3 | 564 |