Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006578.3 | NP_006569.1 | 4, 5 | 459 | ||
NM_016194.3 | NP_057278.2 | 6, 7 | 564 | |||
XM_011521162.2 | XP_011519464.1 | 4, 5 | 816 | |||
XM_011521163.2 | XP_011519465.1 | 4, 5 | 341 | |||
GenBank mRNA | AF017656.1 | - | 4, 5 | 472 | ||
AF300650.1 | - | 5, 6 | 499 | |||
AF501885.1 | - | 4, 5 | 373 | |||
AK290330.1 | - | 4, 5 | 467 | |||
AK314775.1 | - | 4, 5 | 417 | |||
BC013997.2 | - | 4, 5 | 456 | 3621195 |