Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007257.5 | NP_009188.1 | 3 | 1526 | ||
XM_011544365.2 | XP_011542667.1 | 3 | 1625 | |||
GenBank mRNA | AB020690.1 | - | 1 | 933 | ||
AF083114.1 | - | 1 | 112 | |||
AF083115.1 | - | 1 | 514 | |||
AK055325.1 | - | 3 | 1468 | |||
BC036489.1 | - | 3 | 1456 | 5262276 | ||
BC047515.1 | - | 3 | 1431 | 5294654 | ||
BC062301.1 | - | 1 | 1007 | 6580976 |