Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRG3 SIRNA Assays ›
Gene Symbol:
NRG3
Gene Name:
neuregulin 3
Gene Aliases:
HRG3, pro-NRG3
Chromosome Location:
Chr.10: 81875298 - 82989979 on Build GRCh38
UniGene ID:
Hs.125119
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s21055
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRG3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001010848.3
NP_001010848.2
2
862
NM_001165972.1
NP_001159444.1
2
862
NM_001165973.1
NP_001159445.1
3
446
XM_005269444.4
XP_005269501.1
2
863
XM_011539172.2
XP_011537474.1
2
865
XM_011539173.2
XP_011537475.1
2
865
XM_011539175.2
XP_011537477.1
2
865
XM_017015573.1
XP_016871062.1
3
945
XM_017015574.1
XP_016871063.1
3
943
XM_017015575.1
XP_016871064.1
3
945
XM_017015576.1
XP_016871065.1
3
943
XM_017015577.1
XP_016871066.1
3
945
XM_017015578.1
XP_016871067.1
3
943
XM_017015579.1
XP_016871068.1
3
944
XM_017015580.1
XP_016871069.1
3
943
XM_017015581.1
XP_016871070.1
2
865
XM_017015582.1
XP_016871071.1
3
215
XM_017015583.1
XP_016871072.1
3
215
XM_017015584.1
XP_016871073.1
3
956
GenBank mRNA
AK122886.1
-
3
613
AK296563.1
-
2
684
BC136811.1
-
2
862
9020801
DQ001411.1
-
2
835
DQ338459.1
-
2
119
DQ857894.1
-
3
448
HM068873.1
-
2
835
HM068874.1
-
2
835
HM068875.1
-
3
172
HM068876.1
-
2
72
HM068877.1
-
2
72
HM068878.1
-
2
72
HM068879.1
-
2
72
HM068880.1
-
2
147
HM068881.1
-
2
147
HM068882.1
-
3
247
HM068883.1
-
3
247
HM068884.1
-
4
325
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
activation of transmembrane receptor protein tyrosine kinase activity
pattern specification process
chemorepulsion involved in interneuron migration from the subpallium to the cortex
intracellular signal transduction
animal organ development
mammary placode formation
negative regulation of neuron migration
receptor binding
growth factor activity
transmembrane receptor protein tyrosine kinase activator activity
receptor tyrosine kinase binding
chemorepellent activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0