Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011531550.2 | XP_011529852.1 | 1 | 313 | ||
GenBank mRNA | AF016492.1 | - | 1 | 307 | ||
BC069441.1 | - | 1 | 307 | 7262335 | ||
BC107059.2 | - | 1 | 307 | 40006084 | ||
BC107060.2 | - | 1 | 307 | 40006085 |