Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207004.1 | NP_001193933.1 | 2 | 737 | ||
NM_053039.1 | NP_444267.1 | 2 | 737 | |||
XM_017008303.1 | XP_016863792.1 | 2 | 807 | |||
GenBank mRNA | AB451456.1 | - | 2 | 735 | ||
AF177272.1 | - | 2 | 737 | |||
AF177273.1 | - | 2 | 737 | |||
AF177274.1 | - | 3 | 386 |
Gene Symbol: UGT2B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011531550.2 | XP_011529852.1 | 2 | 498 | ||
XM_017007660.1 | XP_016863149.1 | 3 | 250 | |||
XM_017007661.1 | XP_016863150.1 | 2 | 128 | |||
GenBank mRNA | AF016492.1 | - | 2 | 744 | ||
BC069441.1 | - | 2 | 744 | 7262335 | ||
BC107059.2 | - | 2 | 744 | 40006084 | ||
BC107060.2 | - | 2 | 744 | 40006085 |