Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012215.3 | NP_036347.1 | 8, 9 | 1644 | ||
XM_017015585.1 | XP_016871074.1 | 9, 10 | 2021 | |||
XM_017015586.1 | XP_016871075.1 | 5, 6 | 808 | |||
GenBank mRNA | AB014579.2 | - | 8, 9 | 1206 | ||
AF036144.2 | - | 8, 9 | 1596 | |||
AF307332.1 | - | 8, 9 | 1596 | |||
BC032591.1 | - | 6, 7 | 1208 | |||
BC047877.1 | - | 8, 9 | 1599 |