Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098527.2 | NP_001091997.2 | 4 | 1377 | ||
NM_001159545.1 | NP_001153017.1 | 3 | 419 | |||
NM_001159546.1 | NP_001153018.1 | 3 | 448 | |||
NM_006605.3 | NP_006596.2 | 1 | 463 | |||
XM_011529826.1 | XP_011528128.1 | 4 | 1581 | |||
XM_011529827.2 | XP_011528129.1 | 5 | 756 | |||
XM_011529828.1 | XP_011528130.1 | 5 | 1491 | |||
XM_011529830.2 | XP_011528132.1 | 5 | 841 | |||
XM_011529832.1 | XP_011528134.1 | 3 | 1467 | |||
XM_017028535.1 | XP_016884024.1 | 4 | 586 | |||
XM_017028536.1 | XP_016884025.1 | 4 | 635 | |||
GenBank mRNA | AJ010231.1 | - | 1 | 463 | ||
BC051910.1 | - | 3 | 419 | 5729832 | ||
BC069827.1 | - | 1 | 384 | 7262662 | ||
CR456563.1 | - | 3 | 409 |