Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098527.2 | NP_001091997.2 | 5 | 1831 | ||
NM_001159545.1 | NP_001153017.1 | 4 | 873 | |||
NM_001159546.1 | NP_001153018.1 | 4 | 902 | |||
NM_006605.3 | NP_006596.2 | 2 | 917 | |||
XM_011529826.1 | XP_011528128.1 | 5 | 2035 | |||
XM_011529827.2 | XP_011528129.1 | 6 | 1210 | |||
XM_011529828.1 | XP_011528130.1 | 6 | 1945 | |||
XM_011529830.2 | XP_011528132.1 | 6 | 1295 | |||
XM_011529832.1 | XP_011528134.1 | 4 | 1921 | |||
XM_017028535.1 | XP_016884024.1 | 5 | 1040 | |||
XM_017028536.1 | XP_016884025.1 | 5 | 1089 | |||
GenBank mRNA | AJ010231.1 | - | 2 | 917 | ||
AK055999.1 | - | 5 | 1831 | |||
BC051910.1 | - | 4 | 873 | 5729832 | ||
BC069737.1 | - | 2 | 839 | 7262638 | ||
BC069827.1 | - | 2 | 838 | 7262662 | ||
CR456563.1 | - | 4 | 863 |
Gene Symbol: RFPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021026.2 | NP_066306.2 | 2 | 921 | ||
XM_011530298.2 | XP_011528600.1 | 4 | 881 | |||
XM_011530299.2 | XP_011528601.1 | 3 | 783 | |||
XM_017028900.1 | XP_016884389.1 | 6 | 1268 | |||
XM_017028901.1 | XP_016884390.1 | 5 | 1202 | |||
XM_017028902.1 | XP_016884391.1 | 10 | 4246 | |||
GenBank mRNA | AJ010228.1 | - | 2 | 921 | ||
AJ010229.1 | - | 2 | 918 | |||
BC104768.1 | - | 2 | 648 | |||
CR456562.2 | - | 2 | 740 |