Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv420h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115018.1 | NP_001108490.1 | 8 | 898 | ||
NM_146177.2 | NP_666289.2 | 8 | 956 | |||
GenBank mRNA | AK137449.1 | - | 8 | 897 | ||
AK154848.1 | - | 8 | 1496 | |||
AY555193.1 | - | 8 | 910 | |||
BC024816.1 | - | 8 | 895 | 5359433 | ||
BC085473.1 | - | 6 | 652 | 6414100 |