Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146298.2 | NP_666410.2 | 1 | 254 |
Gene Symbol: Olfr748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011837.2 | NP_001011837.2 | 1 | 254 |
Set Membership: |
![]() |