Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013752.3 | NP_038780.3 | 11 | 1707 | ||
GenBank mRNA | AB016988.1 | - | 11 | 1629 | ||
AF076687.1 | - | 11 | 1701 | |||
AF092840.1 | - | 11 | 1690 | |||
AK031933.1 | - | 11 | 1693 | |||
AK132302.1 | - | 11 | 1749 | |||
AK134805.1 | - | 11 | 1743 | |||
AK134960.1 | - | 11 | 1735 | |||
AK159790.1 | - | 11 | 1713 | |||
AK160614.1 | - | 11 | 1674 | |||
AK214489.1 | - | 1 | 123 | |||
BC003719.1 | - | 11 | 1716 | |||
BC013190.1 | - | 11 | 1707 | |||
BC044773.2 | - | 11 | 1706 | 5067851 | ||
BC055061.1 | - | 11 | 1699 | 6528890 |