Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177033.3 | NP_796007.1 | 2 | 998 | ||
GenBank mRNA | AB292670.1 | - | 1 | 401 | ||
AK033944.1 | - | 2 | 792 | |||
AK044771.1 | - | 2 | 533 | |||
AK141514.1 | - | 2 | 999 | |||
BC120564.1 | - | 1 | 460 | 40129487 | ||
BC137825.1 | - | 1 | 461 | 8860844 | ||
DQ421811.1 | - | 1 | 464 |