Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177059.3 | NP_796033.2 | 14, 15 | 1894 | ||
GenBank mRNA | AF374459.1 | - | 14, 15 | 1894 | ||
AK046350.1 | - | 14, 15 | 1945 | |||
AK081114.1 | - | 14, 15 | 1940 | |||
AK220367.1 | - | 10, 11 | 1274 | |||
BC132353.1 | - | 14, 15 | 1871 | 40130630 | ||
BC144824.1 | - | 14, 15 | 1864 |