Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004157.2 | NP_001004157.2 | 8, 9 | 1407 | ||
GenBank mRNA | AK133477.1 | - | 8, 9 | 1390 | ||
AK220215.1 | - | 8, 9 | 1391 | |||
BC057600.1 | - | 7, 8 | 1314 |