Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020277.2 | NP_064673.2 | 7, 8 | 1000 | ||
GenBank mRNA | AB039952.1 | - | 6, 7 | 917 | ||
AF228681.1 | - | 7, 8 | 1000 | |||
AJ271092.2 | - | 6, 7 | 913 | |||
AK035197.1 | - | 3, 4 | 530 | |||
AK044908.1 | - | 3, 4 | 615 | |||
AY280364.1 | - | 7, 8 | 964 | |||
AY280365.1 | - | 7, 8 | 964 | |||
BC133712.1 | - | 6, 7 | 917 | 40143371 |