Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NEK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145001.2 | NP_001138473.1 | 9 | 1071 | ||
NM_001166167.1 | NP_001159639.1 | 8 | 853 | |||
NM_001166168.1 | NP_001159640.1 | 8 | 848 | |||
NM_001166169.1 | NP_001159641.1 | 8 | 857 | |||
NM_001166170.1 | NP_001159642.1 | 8 | 855 | |||
NM_001166171.1 | NP_001159643.1 | 9 | 1020 | |||
NM_014397.5 | NP_055212.2 | 8 | 906 | |||
XM_005251664.1 | XP_005251721.1 | 8 | 912 | |||
XM_006716936.2 | XP_006716999.1 | 8 | 917 | |||
XM_017014217.1 | XP_016869706.1 | 9 | 1080 | |||
GenBank mRNA | AB026289.1 | - | 8 | 837 | ||
AF087909.1 | - | 8 | 842 | |||
AK294614.1 | - | 8 | 857 | |||
AK313071.1 | - | 8 | 807 | |||
BC000101.2 | - | 8 | 852 | 3507262 | ||
BC004174.2 | - | 9 | 1024 | 2958695 | ||
BC004209.2 | - | 9 | 1024 | 2958695 | ||
BC012761.2 | - | 8 | 853 | 3640103 | ||
CR457091.1 | - | 7 | 687 | |||
CR542222.1 | - | 7 | 687 |