Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT9 SIRNA Assays ›
Gene Symbol:
SEPT9
Gene Name:
septin 9
Gene Aliases:
AF17q25, MSF, MSF1, NAPB, PNUTL4, SINT1, SeptD1
Chromosome Location:
Chr.17: 77281410 - 77500596 on Build GRCh38
UniGene ID:
Hs.440932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s21223
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113491.1
NP_001106963.1
7
1283
NM_001113492.1
NP_001106964.1
7
1458
NM_001113493.1
NP_001106965.1
6
1444
NM_001113494.1
NP_001106966.1
6
1397
NM_001113495.1
NP_001106967.1
5
1097
NM_001113496.1
NP_001106968.1
5
692
NM_001293695.1
NP_001280624.1
6
1226
NM_001293696.1
NP_001280625.1
5
508
NM_001293697.1
NP_001280626.1
5
721
NM_001293698.1
NP_001280627.1
5
807
NM_006640.4
NP_006631.2
6
1915
XM_005256962.1
XP_005257019.1
5
565
XM_006721643.2
XP_006721706.1
8
1386
XM_006721644.1
XP_006721707.1
5
608
XM_011524204.1
XP_011522506.1
8
1483
XM_011524206.1
XP_011522508.1
6
1203
XM_011524207.1
XP_011522509.1
6
1216
XM_011524208.2
XP_011522510.1
5
474
XM_017024031.1
XP_016879520.1
6
815
XM_017024032.1
XP_016879521.1
5
814
GenBank mRNA
AB023208.1
-
6
1397
AF123052.1
-
6
1878
AF142408.1
-
6
1360
AF189712.2
-
8
1410
AF189713.3
-
7
1253
AJ312319.1
-
6
1444
AJ312320.1
-
5
486
AJ312321.1
-
6
1915
AJ312322.1
-
6
1386
AK022493.1
-
5
474
AK056495.1
-
5
1097
AK290368.1
-
7
1458
AK299828.1
-
5
721
AK300270.1
-
6
1185
AK303449.1
-
5
501
AK304143.1
-
6
1298
AK316473.1
-
5
807
AL080131.1
-
2
102
BC021192.2
-
6
1138
3346534
BC054004.1
-
8
1113
6146448
BT007215.1
-
6
1103
CN393537.1
-
5
500
DC410238.1
-
5
509
KU178538.1
-
5
458
KU178539.1
-
6
1103
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
protein heterooligomerization
cell division
cell-cell adhesion
positive regulation of nonmotile primary cilium assembly
GTPase activity
protein binding
GTP binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cnw2s:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0