Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: USP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008563.4 | NP_001008563.2 | 18 | 2094 | ||
NM_001110303.3 | NP_001103773.2 | 18 | 1996 | |||
NM_006676.7 | NP_006667.3 | 18 | 1996 | |||
XM_005251665.3 | XP_005251722.1 | 18 | 1984 | |||
XM_005251666.3 | XP_005251723.1 | 13 | 1558 | |||
XM_011518161.2 | XP_011516463.1 | 19 | 2336 | |||
XM_011518162.2 | XP_011516464.1 | 18 | 2515 | |||
XM_017014221.1 | XP_016869710.1 | 13 | 1558 | |||
GenBank mRNA | AB023220.1 | - | 17 | 1914 | ||
AY074877.1 | - | 18 | 2030 | |||
BC039593.1 | - | 18 | 1940 | 5588842 | ||
BQ882312.1 | - | 11 |