Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138964.2 | NP_620414.1 | 2 | 811 | ||
GenBank mRNA | AB084080.1 | - | 2 | 811 | ||
AF506287.1 | - | 2 | 811 | |||
AY089976.1 | - | 2 | 811 | |||
BC100960.2 | - | 2 | 811 | 40004720 | ||
EF577399.1 | - | 2 | 811 |
Set Membership: |
G-protein coupled receptor |