Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYVE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006691.3 | NP_006682.2 | 5 | 1057 | ||
GenBank mRNA | AF118108.1 | - | 5 | 861 | ||
AF127670.2 | - | 5 | 891 | |||
AK075443.1 | - | 5 | 971 | |||
AK300885.1 | - | 3 | 504 | |||
AK312462.1 | - | 5 | 1055 | |||
AY358925.1 | - | 5 | 930 | |||
BC026231.1 | - | 5 | 816 | 4712863 |