Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006701.4 | NP_006692.1 | 1, 2 | 348 | ||
NR_131176.1 | 1, 2 | 348 | ||||
GenBank mRNA | AF023611.1 | - | 1, 2 | 274 | ||
AF023612.1 | - | 1, 2 | 274 | |||
AF146373.1 | - | 1, 2 | 148 | |||
AK314901.1 | - | 1, 2 | 300 | |||
BC001046.2 | - | 1, 2 | 288 | 2823293 | ||
BC019272.1 | - | 1, 2 | 288 | 2823293 | ||
BE385528.1 | - | 1, 2 | 284 | |||
BF972457.1 | - | 1, 2 | 349 | |||
BU553165.1 | - | 292 |