Gene Symbol: TXNL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303471.2 | NP_001290400.1 | 3 | 577 | ||
NM_001305557.1 | NP_001292486.1 | 2 | 404 | |||
NM_001305563.1 | NP_001292492.1 | 3 | 217 | |||
NM_001305564.1 | NP_001292493.1 | 2 | 160 | |||
NM_006701.4 | NP_006692.1 | 2 | 428 | |||
NR_131175.1 | 3 | 577 | ||||
NR_131176.1 | 2 | 428 | ||||
GenBank mRNA | AF023611.1 | - | 2 | 354 | ||
AF023612.1 | - | 2 | 354 | |||
AF146373.1 | - | 2 | 228 | |||
AK308703.1 | - | 2 | 140 | |||
AK309098.1 | - | 3 | 494 | |||
AK314901.1 | - | 2 | 380 | |||
AL522879.3 | - | 3 | 566 | |||
AV729855.2 | - | 2 | 160 | |||
BC001046.2 | - | 2 | 368 | 2823293 | ||
BC019272.1 | - | 2 | 368 | 2823293 | ||
BE385528.1 | - | 2 | 364 | |||
BF972457.1 | - | 2 | 429 | |||
BM922767.1 | - | 2 | 405 | |||
BU553165.1 | - | 372 |