Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006705.3 | NP_006696.1 | 4 | 1005 | ||
XM_011518163.2 | XP_011516465.1 | 3 | 906 | |||
GenBank mRNA | AF079806.1 | - | 4 | 993 | ||
AF087883.1 | - | 4 | 987 | |||
BC000465.2 | - | 4 | 986 | 2822765 | ||
BC019325.1 | - | 4 | 986 | 2823131 | ||
D83023.1 | - | 4 | 978 |