Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000063.5 | NP_000054.2 | 13 | 1952 | ||
NM_001145903.2 | NP_001139375.1 | 11 | 1556 | |||
NM_001178063.2 | NP_001171534.1 | 9 | 1102 | |||
NM_001282457.1 | NP_001269386.1 | 9 | 1159 | |||
NM_001282458.1 | NP_001269387.1 | 13 | 1980 | |||
GenBank mRNA | AK096258.1 | - | 10 | 1257 | ||
AK222537.1 | - | 13 | 1704 | |||
AK222783.1 | - | 13 | 1676 | |||
AK298311.1 | - | 9 | 1102 | |||
AK298808.1 | - | 9 | 1159 | |||
AK300892.1 | - | 11 | 1316 | |||
AK300930.1 | - | 13 | 1718 | |||
AK307930.1 | - | 11 | 1310 | |||
AK312581.1 | - | 13 | 1786 | |||
BC043484.1 | - | 13 | 1687 | 5763774 | ||
BX537504.1 | - | 7 | 1689 | |||
X04481.1 | - | 13 | 1712 |