Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284271.1 | NP_001271200.1 | 3 | 497 | ||
NM_006828.3 | NP_006819.2 | 3 | 497 | |||
NM_022091.4 | NP_071374.1 | 3 | 497 | |||
XM_011535394.2 | XP_011533696.1 | 3 | 507 | |||
XM_011535395.2 | XP_011533697.1 | 3 | 486 | |||
XM_017010205.1 | XP_016865694.1 | 3 | 507 | |||
XM_017010206.1 | XP_016865695.1 | 3 | 489 | |||
GenBank mRNA | AK023189.1 | - | 2 | 83 | ||
AK307903.1 | - | 3 | 426 | |||
AK315197.1 | - | 3 | 431 | |||
AL834463.1 | - | 2 | 182 | |||
BC026066.1 | - | 3 | 494 | |||
BC050681.1 | - | 3 | 398 | |||
BC125211.1 | - | 3 | 256 | |||
BC125212.1 | - | 3 | 256 | |||
CN255848.1 | - | 2 | 146 |