Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284271.1 | NP_001271200.1 | 2 | 412 | ||
NM_006828.3 | NP_006819.2 | 2 | 412 | |||
NM_022091.4 | NP_071374.1 | 2 | 412 | |||
XM_011535394.2 | XP_011533696.1 | 2 | 407 | |||
XM_011535395.2 | XP_011533697.1 | 2 | 401 | |||
XM_011535396.2 | XP_011533698.1 | 2 | 407 | |||
XM_017010205.1 | XP_016865694.1 | 2 | 407 | |||
XM_017010206.1 | XP_016865695.1 | 2 | 404 | |||
GenBank mRNA | AK299117.1 | - | 2 | 412 | ||
AK307903.1 | - | 2 | 341 | |||
AK315197.1 | - | 2 | 346 | |||
AL834463.1 | - | 1 | 97 | |||
BC026066.1 | - | 2 | 409 | |||
BC050681.1 | - | 2 | 313 | |||
BC125211.1 | - | 2 | 171 | |||
BC125212.1 | - | 2 | 171 | |||
CN255848.1 | - | 1 | 61 |