Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297655.1 | NP_001284584.1 | 3 | 310 | ||
NM_001297656.1 | NP_001284585.1 | 2 | 428 | |||
NM_001297657.1 | NP_001284586.1 | 2 | 115 | |||
NM_006845.3 | NP_006836.2 | 3 | 310 | |||
XM_011540540.2 | XP_011538842.1 | 3 | 424 | |||
XM_011540541.2 | XP_011538843.1 | 2 | 365 | |||
GenBank mRNA | AB264115.1 | - | 3 | 310 | ||
AK290688.1 | - | 3 | 236 | |||
AK300756.1 | - | 3 | 267 | |||
AK302267.1 | - | 2 | 83 | |||
AK312606.1 | - | 3 | 266 | |||
AU130795.1 | - | 3 | 310 | |||
AY026505.1 | - | 2 | 381 | |||
BC008764.2 | - | 3 | 249 | 3347571 | ||
BC014924.1 | - | 3 | 247 | 3909438 | ||
BT006759.1 | - | 3 | 195 | |||
CR450302.1 | - | 3 | 195 | |||
DB045012.1 | - | 2 | 60 | |||
DC399082.1 | - | 2 | 83 | |||
DC404550.1 | - | 2 | 410 | |||
U63743.1 | - | 3 | 249 |