Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006851.2 | NP_006842.2 | 3 | 662 | ||
XM_011537775.2 | XP_011536077.1 | 4 | 712 | |||
XM_017018721.1 | XP_016874210.1 | 3 | 542 | |||
GenBank mRNA | AF400440.1 | - | 3 | 542 | ||
AK313449.1 | - | 3 | 528 | |||
BC012510.1 | - | 3 | 578 | 4453841 | ||
EF667354.1 | - | 4 | 585 | |||
U16307.3 | - | 3 | 598 | |||
X91911.1 | - | 3 | 642 |