Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278318.1 | NP_001265247.1 | 7 | 992 | ||
NM_006863.3 | NP_006854.1 | 9 | 1592 | |||
NR_103501.1 | 8 | 1394 | ||||
GenBank mRNA | AF025529.1 | - | 7 | 908 | ||
AF025530.1 | - | 9 | 1520 | |||
AK290919.1 | - | 9 | 1536 | |||
BC093708.1 | - | 8 | 1366 | 7939553 | ||
BC101514.1 | - | 8 | 1366 | 8069020 |
Gene Symbol: LILRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130917.2 | NP_001124389.2 | 7 | 1329 | ||
XM_011526385.1 | XP_011524687.1 | 8 | 1901 | |||
XM_011526388.1 | XP_011524690.1 | 7 | 1865 | |||
GenBank mRNA | AF025531.1 | - | 7 | 1320 | ||
CB046242.1 | - | 2 | 236 | |||
EU915609.1 | - | 6 | 1262 | |||
EU915610.1 | - | 7 | 1298 | |||
U82278.1 | - | 7 | 1281 |