Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4BPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000715.3 | NP_000706.1 | 4 | 599 | ||
XM_005273251.1 | XP_005273308.1 | 4 | 555 | |||
XM_005273252.4 | XP_005273309.1 | 4 | 731 | |||
GenBank mRNA | AK303793.1 | - | 5 | 410 | ||
AK303798.1 | - | 4 | 549 | |||
AK313164.1 | - | 4 | 545 | |||
BC022312.1 | - | 4 | 599 | 4688428 | ||
M31452.1 | - | 4 | 543 | |||
X02865.1 | - | 2 | 168 |