Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017051.2 | NP_058747.1 | 5 | 705 | ||
GenBank mRNA | BC070913.1 | - | 5 | 705 | 7104698 | |
Y00497.1 | - | 5 | 633 |