Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013046.3 | NP_037178.1 | 3 | 395 | ||
XM_006236899.2 | XP_006236961.1 | 3 | 826 | |||
GenBank mRNA | BC161830.1 | - | 3 | 342 | 9091055 | |
FQ100942.1 | - | 3 | 395 | |||
FQ221570.1 | - | 3 | 371 | |||
FQ221573.1 | - | 3 | 371 | |||
FQ222015.1 | - | 3 | 371 | |||
FQ222264.1 | - | 3 | 369 | |||
FQ222377.1 | - | 3 | 370 | |||
FQ222571.1 | - | 3 | 369 | |||
FQ222984.1 | - | 3 | 369 | |||
FQ223091.1 | - | 3 | 371 | |||
FQ228499.1 | - | 3 | 371 | |||
FQ228758.1 | - | 3 | 371 | |||
FQ228762.1 | - | 3 | 369 | |||
FQ229082.1 | - | 3 | 371 | |||
FQ229292.1 | - | 3 | 369 | |||
M12138.1 | - | 3 | 362 | |||
M36317.1 | - | 3 | 362 |