Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031579.2 | NP_113767.1 | 2 | 803 | ||
XM_006244670.2 | XP_006244732.1 | 2 | 685 | |||
XM_006244671.2 | XP_006244733.1 | 2 | 668 | |||
GenBank mRNA | AY062269.1 | - | 1 | 86 | ||
BC081772.1 | - | 1 | 433 | 7124031 | ||
BC097307.1 | - | 2 | 803 | 7446277 | ||
L27843.1 | - | 2 | 632 |
Gene Symbol: LOC103693189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766779.1 | XP_008765001.1 | 1 | 602 |