Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BTN3A2 SIRNA Assays ›
Gene Symbol:
BTN3A2
Gene Name:
butyrophilin subfamily 3 member A2
Gene Aliases:
BT3.2, BTF4, BTN3.2, CD277
Chromosome Location:
Chr.6: 26365159 - 26451697 on Build GRCh38
UniGene ID:
Hs.376046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s21920
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BTN3A2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001197246.2
NP_001184175.1
3
465
NM_001197247.2
NP_001184176.1
5
730
NM_001197248.2
NP_001184177.1
3
569
NM_001197249.2
NP_001184178.1
4
630
NM_007047.4
NP_008978.2
5
720
XM_005248827.3
XP_005248884.1
5
708
XM_005248831.3
XP_005248888.1
5
708
XM_005248832.3
XP_005248889.1
5
708
XM_006714979.3
XP_006715042.1
5
708
XM_006714980.3
XP_006715043.1
5
708
XM_006714981.3
XP_006715044.1
5
698
XM_006714982.3
XP_006715045.1
4
618
XM_011514267.2
XP_011512569.1
5
574
XM_011514268.2
XP_011512570.1
5
708
XM_011514269.2
XP_011512571.1
4
477
XM_017010211.1
XP_016865700.1
5
698
XM_017010212.1
XP_016865701.1
5
574
XM_017010213.1
XP_016865702.1
4
580
XM_017010214.1
XP_016865703.1
4
618
XM_017010216.1
XP_016865705.1
4
618
GenBank mRNA
AF257505.1
-
3
467
AK290880.1
-
5
658
AK299417.1
-
4
514
AK312847.1
-
5
652
BC002832.2
-
5
611
3637548
BC020214.1
-
5
611
4870774
U90144.1
-
4
527
U90546.1
-
5
648
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
T cell mediated immunity
interferon-gamma secretion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f4tdd:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0