Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281444.1 | NP_001268373.1 | 6 | 1201 | ||
NM_001281445.1 | NP_001268374.1 | 4 | 764 | |||
NM_007066.4 | NP_008997.1 | 4 | 779 | |||
NM_181804.2 | NP_861520.1 | 5 | 926 | |||
NM_181805.2 | NP_861521.1 | 6 | 1031 | |||
XM_017027614.1 | XP_016883103.1 | 5 | 910 | |||
XM_017027615.1 | XP_016883104.1 | 4 | 798 | |||
XM_017027616.1 | XP_016883105.1 | 4 | 716 | |||
GenBank mRNA | AB019517.1 | - | 2 | 492 | ||
AF115966.1 | - | 2 | 741 | |||
AF182032.1 | - | 4 | 633 |