Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163014.1 | NP_001156486.1 | 3 | 305 | ||
GenBank mRNA | AK079424.1 | - | 3 | 282 | ||
BC138170.1 | - | 3 | 289 | 8861189 | ||
BC145172.1 | - | 3 | 289 | 9053723 |