Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BVES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199563.1 | NP_001186492.1 | 6 | 974 | ||
NM_007073.4 | NP_009004.2 | 6 | 844 | |||
NM_147147.3 | NP_671488.1 | 6 | 914 | |||
XM_011535398.2 | XP_011533700.1 | 6 | 849 | |||
GenBank mRNA | AF204172.1 | - | 6 | 868 | ||
AK289979.1 | - | 6 | 958 | |||
BC034425.1 | - | 6 | 914 | 4821891 | ||
BC040502.1 | - | 6 | 974 | 5295907 |