Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022902.4 | NP_075053.2 | 1, 2 | 394 | ||
NM_024055.4 | NP_076960.1 | 1, 2 | 394 | |||
XM_006714672.3 | XP_006714735.1 | 1, 2 | 388 | |||
GenBank mRNA | AF461760.1 | - | 1, 2 | 288 | ||
AK292925.1 | - | 1, 2 | 258 | |||
AY089991.1 | - | 1, 2 | 87 | |||
AY358505.1 | - | 1, 2 | 305 | |||
BC000808.1 | - | 1, 2 | 239 | |||
BC017441.1 | - | 1, 2 | 352 | |||
BC130452.1 | - | 1, 2 | 145 | |||
BC130454.1 | - | 1, 2 | 145 | |||
BX537394.1 | - | 1, 2 | 87 | |||
DB090660.1 | - | 1, 2 | 394 | |||
DB259372.1 | - | 1, 2 | 388 |