Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RABL2B SIRNA Assays ›
Gene Symbol:
RABL2B
RABL2A
Gene Name:
RAB, member of RAS oncogene family-like 2B
RAB, member of RAS oncogene family-like 2A
Gene Aliases:
-
-
Chromosome Location:
Chr.22: 50767492 - 50783705 on Build GRCh38
Chromosome Location:
Chr.2: 113627218 - 113643398 on Build GRCh38
UniGene ID:
Hs.584862
Hs.446425
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s22012
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RABL2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003789.1
NP_001003789.1
9
1720
NM_001130919.1
NP_001124391.1
9
1716
NM_001130920.1
NP_001124392.1
10
1875
NM_001130921.1
NP_001124393.1
10
1968
NM_001130922.1
NP_001124394.1
9
1717
NM_001130923.1
NP_001124395.1
10
1902
NM_007081.2
NP_009012.1
9
1713
XM_005261899.2
XP_005261956.1
10
1999
XM_011530668.1
XP_011528970.1
10
2002
XM_011530669.2
XP_011528971.1
9
1749
XM_011530670.1
XP_011528972.1
9
1766
XM_011530671.1
XP_011528973.1
10
1876
XM_011530673.1
XP_011528975.1
10
1990
XM_011530675.1
XP_011528977.1
10
1969
XM_017028543.1
XP_016884032.1
10
2192
XM_017028544.1
XP_016884033.1
10
2097
XM_017028545.1
XP_016884034.1
10
2038
XM_017028546.1
XP_016884035.1
10
2040
XM_017028547.1
XP_016884036.1
10
2189
XM_017028548.1
XP_016884037.1
9
1746
XM_017028549.1
XP_016884038.1
10
2035
XM_017028550.1
XP_016884039.1
10
1864
XM_017028551.1
XP_016884040.1
8
1908
XM_017028552.1
XP_016884041.1
10
2162
XM_017028553.1
XP_016884042.1
10
2064
XM_017028554.1
XP_016884043.1
10
2008
XM_017028555.1
XP_016884044.1
10
2010
XM_017028556.1
XP_016884045.1
10
2159
XM_017028557.1
XP_016884046.1
10
2005
XM_017028558.1
XP_016884047.1
10
1843
GenBank mRNA
AF095352.1
-
9
1660
AK094341.1
-
3
2042
BC014879.2
-
9
1681
3906749
BC024281.1
-
10
1918
4580599
Gene Symbol: RABL2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306158.1
NP_001293087.1
9
1722
NM_001306161.1
NP_001293090.1
7
1534
NM_013412.3
NP_038198.1
9
1719
XM_005263569.3
XP_005263626.1
9
1763
XM_005263570.3
XP_005263627.1
9
1730
XM_006712207.3
XP_006712270.1
9
1832
XM_006712208.3
XP_006712271.1
9
1721
XM_006712209.3
XP_006712272.1
9
1875
XM_006712215.3
XP_006712278.1
10
1932
XM_011510509.2
XP_011508811.1
9
1723
XM_017003203.1
XP_016858692.1
8
2472
XM_017003204.1
XP_016858693.1
9
2412
XM_017003206.1
XP_016858695.1
10
1873
XM_017003207.1
XP_016858696.1
9
1786
XM_017003208.1
XP_016858697.1
9
1743
XM_017003209.1
XP_016858698.1
9
1727
XM_017003210.1
XP_016858699.1
8
2460
XM_017003211.1
XP_016858700.1
9
1751
XM_017003212.1
XP_016858701.1
8
2457
XM_017003213.1
XP_016858702.1
8
2442
XM_017003214.1
XP_016858703.1
9
2382
XM_017003216.1
XP_016858705.1
9
1845
XM_017003217.1
XP_016858706.1
9
1729
XM_017003218.1
XP_016858707.1
10
1883
XM_017003219.1
XP_016858708.1
9
1796
XM_017003220.1
XP_016858709.1
8
2439
XM_017003221.1
XP_016858710.1
9
1715
XM_017003222.1
XP_016858711.1
10
1870
XM_017003223.1
XP_016858712.1
6
2280
XM_017003224.1
XP_016858713.1
7
1547
XM_017003225.1
XP_016858714.1
7
1538
XM_017003226.1
XP_016858715.1
6
2250
XM_017003227.1
XP_016858716.1
7
1517
XM_017003228.1
XP_016858717.1
8
1645
XM_017003229.1
XP_016858718.1
9
1739
XM_017003230.1
XP_016858719.1
9
1753
XM_017003232.1
XP_016858721.1
8
1600
GenBank mRNA
AF095351.1
-
9
1653
AK092971.1
-
3
2451
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
GTPase activity
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-sq56x:80/10.239.161.141:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline