Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053828.1 | NP_446280.1 | 2 | 177 | ||
GenBank mRNA | L26913.1 | - | 2 | 177 |