Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197907.1 | NP_001184836.1 | 11 | 2125 | ||
NM_057153.3 | NP_476494.2 | 9 | 1807 | |||
GenBank mRNA | BC166763.1 | - | 9 | 2023 | 7931847 |