Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM107A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076778.2 | NP_001070246.1 | 4 | 1353 | ||
NM_001282713.1 | NP_001269642.1 | 4 | 1153 | |||
NM_001282714.1 | NP_001269643.1 | 5 | 1601 | |||
NM_007177.3 | NP_009108.1 | 5 | 1451 | |||
GenBank mRNA | AB023811.1 | - | 4 | 934 | ||
AF035283.1 | - | 4 | 1352 | |||
AF089853.1 | - | 4 | 1444 | |||
AF089854.1 | - | 5 | 1542 | |||
AK054720.1 | - | 5 | 1601 | |||
AK055443.1 | - | 3 | 1489 | |||
AK128791.1 | - | 4 | 980 | |||
AK298001.1 | - | 4 | 1108 | |||
AK316450.1 | - | 4 | 1008 | |||
AL050264.1 | - | 4 | 966 | |||
BC010561.1 | - | 5 | 1052 | 4152118 | ||
EU569833.1 | - | 4 | 1008 |