Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM107A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076778.2 | NP_001070246.1 | 4 | 1668 | ||
NM_001282713.1 | NP_001269642.1 | 4 | 1468 | |||
NM_001282714.1 | NP_001269643.1 | 5 | 1916 | |||
NM_007177.3 | NP_009108.1 | 5 | 1766 | |||
GenBank mRNA | AF035283.1 | - | 4 | 1667 | ||
AF089853.1 | - | 4 | 1757 | |||
AF089854.1 | - | 5 | 1855 | |||
AK054720.1 | - | 5 | 1916 | |||
AK055443.1 | - | 3 | 1804 | |||
AK128791.1 | - | 4 | 1295 | |||
AK316450.1 | - | 4 | 1323 | |||
AL050264.1 | - | 4 | 1281 | |||
BC010561.1 | - | 5 | 1367 | 4152118 | ||
EU569833.1 | - | 4 | 1323 |