Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013863.2 | NP_001013885.2 | 5 | 1198 | ||
XM_008768859.1 | XP_008767081.1 | 6 | 1231 | |||
XM_008768860.1 | XP_008767082.1 | 7 | 1647 | |||
XM_008768861.1 | XP_008767083.1 | 7 | 1396 | |||
GenBank mRNA | BC086565.1 | - | 5 | 1348 | 7113479 |