Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134413.1 | NP_001127885.1 | 5 | 989 | ||
GenBank mRNA | BC158884.1 | - | 5 | 989 | 7938728 |